DWK Life Sciences Wheaton™ E-Z Seals™ Aluminum Vial Caps
Lined seals for use with crimp-top vials
Supplier: DWK Life Sciences 22421905
Description
- Ideal closures for chromatography and other instrumentation applications
- Open top seal provides for filling or retrieving contents with a syringe
Specifications
PTFE/Silicone | |
Aluminum | |
1000 Cs. | |
11 mm |
Blue | |
E-Z Seal | |
Open Top Cap |