Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials with Rubber-Lined Caps
Manufacturer:  Wheaton™ 224721
 View more versions of this product
Catalog No. 03-343-3B
Specifications Description & Specifications
Specifications
Yes
Screw cap
12mm
Clear Low Extractable Borosilicate Glass
2mL
38mm
14B Rubber
Screw
USP Type I, ASTM E438, Type I, Class A
2.0mL
Clear
E-C Sample Vial
8-425
Phenolic
GPI 8-425
0.5 dr.