Please login to your online account to display your discounted pricing
Wheaton™ Dry Seal™ Desiccator
No need to use flange grease
$824.71 - $824.71
Specifications
Material Soda Lime Glass
Product Type Dry-seal Desiccator
Quantity Each
Includes Lift-off-cover, 40/35 standard taper joint sleeve valve, silicone O-ring and aluminum plate
Max. Vacuum 914hPa
View More Specs
Includes:  Cover, sleeve valve, silicone O-ring, and aluminum plate
Products
Catalog Number Mfr. No. Diameter (English)Top Diameter (Metric) Plate Diameter (Metric)Top Price Quantity    
02-913-361
wheaton™
W365887
9.84 in. 23.4cm 25cm Case for $824.71
Description & Specifications
Specifications
Material Soda Lime Glass
Product Type Dry-seal Desiccator
Quantity Each
Includes Lift-off-cover, 40/35 standard taper joint sleeve valve, silicone O-ring and aluminum plate
Max. Vacuum 914hPa
Temperature Range (English) 59° to 176°F
Temperature Range (Metric) 15° to 80°C
  • Silicone O-ring fits into grooves in both flanges
  • Forms an airtight seal as lid is compressed onto lower body by vacuum
  • As soon as vacuum is released, you can lift up lid; no need to twist or slide it open