missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Disposable™ 300/350μL Pipette Microtips
Premium pipette tips covering most research and routine applications, compatible with most pipettes
$1,027.65 - $1,089.03
Specifications
Clean Claims | DNase, RNase Free |
---|---|
Color | Natural |
Material | Virgin PP (Polypropylene) |
Volume (Metric) | 300 μL |
For Use With (Application) | Premium pipette tips covering most research and routine applications, compatible with most pipettes |
Description
- For both fixed- and variable-volume models
- Precision injection molding care for air-tight fit
- Made from high-grade, virgin polypropylene
- Free from trace metals for excellent accuracy and reproducibility
Specifications
DNase, RNase Free | |
Virgin PP (Polypropylene) | |
Premium pipette tips covering most research and routine applications, compatible with most pipettes |
Natural | |
300 μL |