DWK Life Sciences Wheaton™ Disposable™ 300/350μL Pipette Microtips
Premium pipette tips covering most research and routine applications, compatible with most pipettes
Supplier: DWK Life Sciences 85118104
Description
- For both fixed- and variable-volume models
- Precision injection molding care for air-tight fit
- Made from high-grade, virgin polypropylene
- Free from trace metals for excellent accuracy and reproducibility
Specifications
300 μL | |
Sterile | |
DNase, RNase Free | |
Rack | |
Single-use | |
Polypropylene |
Filtered | |
Natural | |
Premium pipette tips covering most research and routine applications, compatible with most pipettes | |
Micro Point | |
Wheaton |