Please login to your online account to display your discounted pricing

Wheaton™ Disposable 2, 5 and 10mL Pipette Macrotips

Premium pipette tips covering most research and routine applications, compatible with most pipettes

$190.20 - $321.40

Specifications

Certifications/Compliance DNase, RNase, pyrogen free
Autoclavable Yes
For Use With (Equipment) Various pipette brands
Material Polypropylene
Packaging Bulk
View More Specs
Products
Catalog Number Mfr. No. Volume (Metric) Price Quantity    

21-263-1B

 
wheaton™
851355
2mL Case of 250 Each for $321.40

21-263-1D

 
wheaton™
851358
10mL Case of 100 Each for $190.20
Description & Specifications

Specifications

Certifications/Compliance DNase, RNase, pyrogen free
Autoclavable Yes
For Use With (Equipment) Various pipette brands
Material Polypropylene
Packaging Bulk
Product Type MacroTip
Sterility Nonsterile
Color Natural
Tip Style Disposable

  • For both fixed- and variable-volume models
  • Precision injection molding care for air-tight fit
  • Made from high-grade, virgin polypropylene
  • Free from trace metals for excellent accuracy and reproducibility
  • Bulk packed

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.