DWK Life Sciences Wheaton™ Disposable 1000μL Pipette Microtips
Premium pipette tips covering most research and routine applications, compatible with most pipettes
$1,013.50 - $1,539.32
Specifications
Clean Claims DNase, RNase Free
Material Virgin PP (Polypropylene)
Volume (Metric) 1 to 1000 μL
For Use With (Application) Premium pipette tips covering most research and routine applications, compatible with most pipettes, excellent accuracy and reproducibility
View More Specs
Products 2
Catalog Number Mfr. No. Color Sterility Format Price Quantity  
Catalog Number Mfr. No. Color Sterility Format Price Quantity  
03-400-12
View Documents Promotion Details
DWK Life Sciences
85118001
Natural Sterile Individually Wrapped
Case of 1000 Each for $1,539.32
03-400-13
View Documents Promotion Details
DWK Life Sciences
85118002
Natural Sterile Rack
Case of 1000 Each for $1,013.50
Description
Description
  • For both fixed- and variable-volume models
  • Precision injection molding care for air-tight fit
  • Made from high-grade, virgin polypropylene
  • Free from trace metals for excellent accuracy and reproducibility
Specifications
Specifications
DNase, RNase Free
Virgin PP (Polypropylene)
1 to 1000 μL
Premium pipette tips covering most research and routine applications, compatible with most pipettes, excellent accuracy and reproducibility