DWK Life Sciences Disposable 10μL Pipette Microtips
Premium pipette tips covering most research and routine applications, compatible with most pipettes
Supplier: DWK Life Sciences W85123901
Description
- For both fixed- and variable-volume models
- Precision injection molding care for air-tight fit
- Made from high-grade, virgin polypropylene
- Free from trace metals for excellent accuracy and reproducibility
Specifications
10 μL | |
Natural | |
Premium pipette tips covering most research and routine applications, compatible with most pipettes, excellent accuracy and reproducibility | |
Standard Length | |
Single-use |
Non-sterile | |
Autoclavable | |
Rack | |
960 | |
Virgin Polypropylene |