missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ CryoFILE™ Tissue Vial Storage Boxes

Storage Boxes for batching and storing Wheaton CryoELITE Tissue Vials (Cat. No. 13-709-140)

Manufacturer:  DWK Life Sciences WW651610Y

 View more versions of this product

Catalog No. 13-709-142



Description

Description

CryoFILE partitions are numbered from 1-25 for easy content identification, with the numbering system printed on lid and bottom of box. Available in six colors, to provide easy sample identification. Made of water-repellent material for longer durability, the CryoFILE Tissue Storage Boxes are able to withstand cryogenic temperatures to -196°C.

  • Holds 25 tissue vials
  • Used for batching and storing cryogenic tissue vials
  • Two boxes can be stacked in a standard 2 in. freezer rack, allowing for storage of 50 vials in standard racks
  • Partitions numbered 1-25 for easy content identification
  • Water repellent material allows for longer durability
  • Dimensions: 130 x 130 x 25mm

Specifications

Specifications

Yellow
25
-384.8°F
50 vials
Tissue Vial Storage Box
-196°C
Documents