missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ CryoFile™ and CryoFile XL™ Storage Boxes

For easy Identification and storage of cryogenic vials

Manufacturer:  DWK Life Sciences WW651604XL

 View more versions of this product

Catalog No. 13-709-109



Description

Description

  • Accommodates 1.2 - 2mL vials and XL boxes store 5 mL Partitioned carton numbered from 1-81 keeps vials organized and easy to identify
  • Numbering system printed on box lid and bottom of box provides easy indexing
  • Water repellent allow for longer durability
  • Withstands cryogenic temperatures to -196°C

Specifications

Specifications

19.18 x 15.87 x 7.25 in. (48.7 x 40.3 x 18.4cm)
3, 4 and 5mL vial
Corrugated Chipboard
XL
9.7cm
13cm
13cm
For easy identification and storage of cryogenic vials
1 to 81 vials
CryoFile™ Storage Box
Blue
3.8 in.
5.1 in.
5.1 in.
Documents