missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CryoELITE™ Cryogenic Vials, Round Bottom with Internal Screw Cap

Internal threaded screw cap can be easily removed with one hand

Manufacturer:  Dwk Life Sciences Inc WW985900

 View more versions of this product

Catalog No. 02-912-701


 

Specifications Description & Specifications

Specifications

Natural
DOT, IATA
0.47 in.
12mm
Yes
No
49mm
Round
Cryogenic Vial
Screw
Natural; With cap and patch
2mL
Polypropylene
Internal
1.92 in.
1000/Case
Yes