missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ CryoELITE™ Cryogenic Vials, Round Bottom with Internal Screw Cap

Internal threaded screw cap can be easily removed with one hand

Manufacturer:  DWK Life Sciences WW985912

 View more versions of this product

Catalog No. 02-912-707



Description

Description

  • Exacting round bottom cryogenic vial allows for up to 17,000 × g max. rcf
  • Unrivaled cap seal exceeds DOT and IATA regulations, ensuring ultimate protection of samples during transportation and demanding freeze-thaw handling
  • Lot certified RNase/DNase and Endotoxin free, sterile and non-pyrogenic provides assurance for sample integrity
  • Made from low binding cryogenic, virgin polypropylene

Specifications

Specifications

Natural
DOT, IATA
Natural
3mL
Polypropylene
Internal
63mm
500/Case
Yes
Sterile
Screw
With Writing Patch
0.47 in.
12mm
Yes
2.48 in.
Marking Spot
Round
Cryogenic Vial
Documents
Accessories