Wheaton™ Crimpenstein™ Electric Crimper/Decapper - Standard Voltage
Lightweight and easy to operate
$2,504.23 - $2,504.23
Specifications
Material Aluminium
View More Specs
Includes: Each Crimpenstein comes with a detachable power cord, a convenient carrying/storage case, a crimper and deapper jaw set or an individual crimper or decapper jaw.
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With Type Price Quantity  
Catalog Number Mfr. No. For Use With Type Price Quantity  
029110011 Wheaton™
W225602-A
13mm Aluminum Seals Crimpenstein with 13mm Crimper and Decapper Jaw-Set Each for $2,504.23
Specifications
Description
Wheaton Crimpenstein Electric Crimper/Decapper
  • Easily attaches and removes aluminum seals on crimp top/serum finish bottles and vials to improve productivity
  • Adjustable jaw intensity with memory allows user to regulate crimper and decapper jaw action
  • Quick-change jaw-sets available in 11, 13 and 20mm sizes allow for crimping and decapping of different size bottles and vials
  • Lightweight and easy to operate enables semiautomatic operation for extended periods with less effort
  • Electric powered for quick setup and high volume operation
For information on using the Crimpenstein with flip cap aluminum seals, visit www.wheatonsci.com/crimp. For information on Wheaton crimp top/serum finish bottles and vials, aluminum seals and rubber stoppers, visit www.fishersci.com.
Optional sturdy stand (Part No. W225630) available.
Order Info
Crimper and Decapper Jaws (Part No. W225616-W225628) and sturdy stand (Part No. W225630) are sold separately. For Crimper Decappers with non-standard voltages, see cat. no. 12912828.
Description & Specifications
Specifications
Aluminium