DWK Life Sciences Wheaton™ Crimpenstein™ Crimper and Decapper Jaws
Quick-change crimper and decapper jaws
$363.60 - $363.60
Specifications
For Use With (Equipment) Crimpenstein Electric Crimper/Decapper
View More Specs
Products 1
Catalog Number Mfr. No. Closure Type For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Closure Type For Use With (Application) Price Quantity  
029110020
View Documents Promotion Details
DWK Life Sciences
225623
20 mm Standard aluminum seals
Case for $363.60
Description
Description
Expand the utilization of the Crimpenstein Electric Crimper/Decapper by purchasing a different size Crimper and/or Decapper Jaw. Jaws are available in a “JawSet” or an individual Crimper or Decapper Jaw in 11, 13 and 20mm sizes.
Specifications
Specifications
Crimpenstein Electric Crimper/Decapper