Please login to your online account to display your discounted pricing

Wheaton™ Complete Cyanide Distillation Apparatus: Components


Components for use with Wheaton Complete Cyanide Distillation Apparatus (cat. no. 09108)

Manufacturer: wheaton™  377161

 View more versions of this product

Catalog No. 09-108-1

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Cyanide distillation kit
Product Type Absorber Jacket