missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ Clear Straight-Sided Jars without Caps

Ideal for soil sampling and environmental applications

Manufacturer:  Wheaton™ W216919

 View more versions of this product

Catalog No. 02-912-190


 

Specifications

Description

Jar; Wheaton; Clear; Straight-Sided; Wide Mouth; Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass; Capacity: 2 oz. (60mL)

Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass

  • Wide mouths provide easy access to contents when using spoons or spatulas
  • Chemical-resistant caps conform to EPA and FDA regulations
  • Available in partitioned Convenience or Valu-Bulk™ cartons
  • Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper
  • Evident labels to protect sample integrity in transit or long-term storage

Description & Specifications

Specifications

2 oz., 60mL
2 oz.
2.16 in.
Soda Lime Glass
55mm
Screw
60mL
Clear
1.8 in.
53-400
48mm