missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps
Ideal for soil sampling and environmental applications
Supplier: DWK Life Sciences W216907
Description
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Wide mouths provide easy access to contents when using spoons or spatulas
Each case of convenience pack bottles includes hazard communication labels for proper content identification and tamper evident labels to protect sample integrity in transit or long-term storage
Order Info
Caps can be ordered separately.
Compliance
Clear-USP Type III soda-lime glass
Specifications
Polypropylene | |
95 mm | |
Screw Cap | |
89-400 mm | |
1000 mL | |
Clear | |
Glass | |
White |
170 mm | |
6.69 in. | |
Poly/Vinyl | |
32 oz., 1000 mL | |
32 oz. | |
3.74 in. | |
Convenience pack |