missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps

Ideal for soil sampling and environmental applications

Manufacturer:  DWK Life Sciences W216906

 View more versions of this product

Catalog No. 02-911-409



Description

Description

Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass

  • Wide mouths provide easy access to contents when using spoons or spatulas

Each case of convenience pack bottles includes hazard communication labels for proper content identification and tamper evident labels to protect sample integrity in transit or long-term storage

Order Info

Caps can be ordered separately.

Compliance

Clear-USP Type III soda-lime glass

Specifications

Specifications

16 oz., 500mL
16 oz.
Poly/Vinyl
3.58 in.
Glass
White
91mm
Screw
500mL
Polypropylene
Clear
3.7 in.
Convenience pack
89-400
95mm
Documents