DWK Life Sciences Wheaton™ Clear Standard Wide-Mouth Bottles: With Polyvinyl-Lined Caps
Ideal for the storage of dry materials and specimens
$60.83 - $128.20
Specifications
Closure Liner Poly/Vinyl
Closure Material Polypropylene
Closure Type Screw Cap
Closure Color White
Closure Size 63-400 mm
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-911-422
View Documents
DWK Life Sciences
W216924
125 mL
Case of 24 Each for $64.66
02-911-423
View Documents
DWK Life Sciences
W216925
250 mL
Case of 24 Each for $87.14
02-911-424
View Documents
DWK Life Sciences
W216926
500 mL
Case of 24 Each for $128.20
02-911-425
View Documents
DWK Life Sciences
W216927
2000 mL
Case of 6 Each for $73.94
02-911-426
View Documents
DWK Life Sciences
W216928
4000 mL
Case of 4 Each for $60.83
Description
Description
  • Clear for easy viewing of contents
  • Wide mouth for easy access to contents
  • Rounded shoulders
Order Info
Available with caps attached or without caps. Caps can be ordered separately.
Compliance
Clear Type III soda-lime glass
Specifications
Specifications
Poly/Vinyl
Screw Cap
63-400 mm
Soda Lime Glass
Polypropylene
White
Clear
SDS