Please login to your online account to display your discounted pricing

Wheaton™ Clear Standard Wide-Mouth Bottles: No Caps


Ideal for the storage of dry materials and specimens

Manufacturer: wheaton™  W216936

 View more versions of this product

Catalog No. 02-912-197

  • / Case of 24 Each
 

Description & Specifications

Specifications

Closure Type Screw
Diameter (Metric) 79mm
Closure Size 63-400
Height (Metric) 133mm
Diameter (English) 3.11 in.
Material Soda Lime Glass
Capacity (English) 16 oz.
Height (English) 5.23 in.
Color Clear
Capacity (Metric) 500mL
Capacity 16 oz., 500mL
Closure Material PTFE
Accessories

Accessories for Catalog Number: 02912197

Item Mfr. No. Item Description Price Per Unit Quantity  
02-911-233
Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners
wheaton™
239243
Excellent for environmental samples, pharmaceuticals and diagnostic reagents Case of 48 Each for $270.29