missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Clear Boston Rounds Without Caps

Manufacturer:  DWK Life Sciences W216834

 View more versions of this product

Catalog No. 02-912-162


 

Description

Description

  • Clear Type III soda-lime glass
Specifications

Specifications

16.9 oz., 500mL
16.9 oz.
Poly/Vinyl
2.95 in.
Boston Round Bottles
28-400
168mm
500mL
Polypropylene
Clear
6.6 in.
White
75mm
Screw
Documents