Please login to your online account to display your discounted pricing
Wheaton™ Clear Boston Rounds with TFE-lined Caps
Ideal for solvent, chemical and sample storage
$43.74 - $108.29
Specifications
Certifications/Compliance EPA, FDA
Closure Liner Polyethylene
Closure Material Polypropylene
Closure Screw
Closure Color White
View More Specs
Products
Catalog Number Mfr. No. Capacity Height (English) Height (Metric) Material Price Quantity    
02-911-326
wheaton™
W216806
1 oz., 30mL 3.1 in. 79mm Glass Case of 48 Each for $108.29
02-911-327
wheaton™
W216807
2 oz., 60mL 3.7 in. 94mm Soda Lime Glass Case of 24 Each for $64.66
02-911-328
wheaton™
W216808
4 oz., 125mL 4.4 in. 112mm Glass Case of 24 Each for $74.11
02-911-330
wheaton™
W216810
16 oz., 500mL 6.6 in. 168mm Soda Lime Glass Case of 12 Each for $46.77
02-911-331
wheaton™
W216811
32 oz., 1000mL 8.18 in. 208mm Glass Case of 12 Each for $69.40
02-911-329
wheaton™
W216809
8 oz., 250mL 5.43 in. 138mm Glass Case of 12 Each for $43.74
Description & Specifications
Specifications
Certifications/Compliance EPA, FDA
Closure Liner Polyethylene
Closure Material Polypropylene
Closure Screw
Closure Color White
Color Clear
  • Narrow mouth enables liquids to be carefully poured to prevent spillage
Available with caps attached or without caps (order caps separately).
USP Type-III soda-lime glass
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.