missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps

Ideal for solvent, chemical and sample storage

Manufacturer:  DWK Life Sciences W216800

 View more versions of this product

Catalog No. 02-911-320



Description

Description

  • Narrow mouth enables liquids to be carefully poured to prevent spillage
  • Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage

  • Available in partitioned Convenience or Valu-Bulk™ cartons

Order Info

Caps can be ordered separately.

Compliance

USP Type-III soda-lime glass

Specifications

Specifications

1 oz., 30mL
1 oz.
Polypropylene
Clear
3.1 in.
Convenience pack
20-400
79mm
30mL
EPA, FDA
Poly/Vinyl
1.22 in.
Soda Lime Glass
White
31mm
Screw
Documents