missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps
Ideal for solvent, chemical and sample storage
Supplier: DWK Life Sciences W216829
Description
Ideal for solvent, chemical and sample storage
- Narrow mouth enables liquids to be carefully poured to prevent spillage
- Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
- Available in partitioned Convenience or Valu-Bulk™ cartons
Order Info
Caps can be ordered separately.
Compliance
USP Type-III soda-lime glass
Specifications
Polypropylene | |
94 mm | |
Screw Cap | |
33-400 mm | |
1000 mL | |
Clear | |
Flint Glass | |
White |
208 mm | |
8.18 in. | |
Poly/Vinyl | |
32 oz., 1000 mL | |
32 oz. | |
3.7 in. | |
Valu-bulk pack | |
EPA, FDA |