missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ Clear Boston Rounds with 14B White Rubber-Lined Caps

Clear glass bottles with caps

Manufacturer:  Wheaton™ W216821

 View more versions of this product

Catalog No. 02-911-341


 

Specifications Description & Specifications

Specifications

8 oz., 250mL
8 oz.
Phenolic
Clear
5.43 in.
Black
60mm
Screw
250mL
USP Type III
14B White Rubber
2.36 in.
Soda Lime Glass
24-400
138mm