missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask
Membrane cell culture flasks ideal for antibody and protein production
Supplier: DWK Life Sciences WCL10003
Includes: User manual with feeding and harvesting schedules
Description
High-density cell culture flasks with dual membrane system allow for less culture maintenance and a high concentration harvest.
- Bulk media chamber allows for longer run times without media refreshing
- Cell compartment concentrates cells, antibodies and proteins for simplified purification
- For suspension cell culture and adherent cell culture: Hybridoma, CHO, NS0, SF, BHK, HEK, etc.
02-912-745, 02-912-742
- For adherent cell cultivation
- 1000 mL media compartment
- 15 mL cell compartment
- Available as package of one (02-912-745) or package three (02-912-742)
- For suspension cell cultivation
- 350 mL media compartment
- 5 mL cell compartment
- Available as package of one (02-912-746) or package five (02-912-746)
- For suspension cell cultivation
- 1000 mL media compartment
- 15 mL cell compartment
- Available as package of one (02-912-744) or package three (02-912-741)
Specifications
0.527 oz. | |
15 mL | |
Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures | |
3/Cs. |
35.2 oz. | |
1000 mL | |
User manual with feeding and harvesting schedules | |
Sterile |