Please login to your online account to display your discounted pricing

Wheaton™ CELLine™ Bioreactor Flask


Membrane cell culture flasks ideal for antibody and protein production

Manufacturer: wheaton™  WWCL1000CHO3L

 View more versions of this product

Catalog No. 02-912-662

  • / Case
 

Description & Specifications

Specifications

Capacity (English) Cell 0.527 oz.
Capacity (English) Media 35.195 oz.
Capacity (Metric) Cell 15mL
Capacity (Metric) Media 1000mL
For Use With (Application) Membrane cell culture flasks ideal for antibody and Protein production
Includes 3L BalanCD CHO growth A media, User manual with feeding and harvesting schedules
Sterile Yes