Please login to your online account to display your discounted pricing

Wheaton™ CELLine™ Bioreactor Flask


Membrane cell culture flasks ideal for antibody and protein production

Manufacturer: wheaton™  WWCL0350CHO1L

 View more versions of this product

Catalog No. 02-912-661

  • / Case
 

Description & Specifications

Specifications

Capacity (English) Cell 0.175 oz.
Capacity (English) Media 12.3 oz.
Capacity (Metric) Cell 5mL
Capacity (Metric) Media 350mL
For Use With (Application) Membrane cell culture flasks ideal for antibody and Protein production
Includes 1L BalanCD CHO growth A media, User manual with feeding and harvesting schedules
Sterile Yes