Please login to your online account to display your discounted pricing

Wheaton™ CELLine™ Bioreactor Flask


Membrane cell culture flasks ideal for antibody and protein production

Manufacturer: wheaton™  WCL1000-1

 View more versions of this product

Catalog No. 02-912-744

  • / Pack
 

Description & Specifications

Specifications

Capacity (English) Media 35.195 oz.
Capacity (Metric) Media 1000mL
Capacity (Metric) Cell 15mL
Includes User manual with feeding and harvesting schedules
Sterile Yes
Capacity (English) Cell 0.527 oz.
For Use With (Application) Membrane cell culture flasks ideal for antibody and Protein production, for suspension cultures