Please login to your online account to display your discounted pricing

Wheaton™ Bottom-Drive Standard Roller Apparatus


For anchorage dependent cell growth in roller bottles

Manufacturer: wheaton™  WWSBPR5031A

 View more versions of this product

Catalog No. 02-924-923

  • / Each
 

Description & Specifications

Specifications

Capacity (English) 15 Bottle positions, Options
For Use With (Application) For laboratory, pilot plant and production cell culture installations
Quantity 3
Spacing 6 in.
Format 6 in.