Please login to your online account to display your discounted pricing

Wheaton™ Bottom-Drive Standard Roller Apparatus


For anchorage dependent cell growth in roller bottles

Manufacturer: wheaton™  WWSBPR5010A

 View more versions of this product

Catalog No. 02-924-920

  • / Each
 

Description & Specifications

Specifications

Format 6 in.
Quantity 1
For Use With (Application) For laboratory, pilot plant and production cell culture installations
Capacity (English) 5 Bottle positions
Spacing 6 in.