Please login to your online account to display your discounted pricing

Wheaton™ Bottom-Drive Standard Roller Apparatus


For anchorage dependent cell growth in roller bottles

Manufacturer: wheaton™  WWSBMR5091A

 View more versions of this product

Catalog No. 02-924-919

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Format 7 in.
Capacity (English) 45 Bottle positions, Options
Spacing 7 in.
For Use With (Application) For laboratory, pilot plant and production cell culture installations
Quantity 9