DWK Life Sciences Wheaton™ Bottom-Drive R2P 2.0 Roller Apparatus
Advanced Production Scale Roller Culture Equipment
Supplier: DWK Life Sciences WRBPR5031A
Description
- Color touch screen or digital display
- Icon driven interface with intuitive menus
- Surface mount gold plated electrical connections for better reliability in high temperature environments
- Updated industry standard networking communications protocols and connections
- Lower power consumption for longer battery run time and safer design
- Backwards compatibility for continuing support of existing installations
- Retrofit kits available to update older Wheaton systems or unsupported systems
- Maintenance free brushless DC motors for years of trouble-free operation
Specifications
120 V | |
Bottom Drive Roller Culture System with options and 3 deck | |
15, Options | |
6 in. |
Bottles | |
For laboratory, pilot plant and production cell culture installations | |
3/Pk. |