Please login to your online account to display your discounted pricing

Wheaton™ Bottom-Drive R2P 2.0 Roller Apparatus


Advanced Production Scale Roller Culture Equipment

Manufacturer: wheaton™  WWRBPR5101A

 View more versions of this product

Catalog No. 02-924-947

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Capacity (English) 50 Bottle positions, Options
For Use With (Application) For laboratory, pilot plant and production cell culture installations
Quantity 10
Spacing 6 in.
Format 6 in.