Please login to your online account to display your discounted pricing

Wheaton™ Bottom-Drive R2P 2.0 Roller Apparatus


Advanced Production Scale Roller Culture Equipment

Manufacturer: wheaton™  WWRBMR5031A

 View more versions of this product

Catalog No. 02-924-955

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Capacity (English) 15 Bottle positions, Options
For Use With (Application) For laboratory, pilot plant and production cell culture installations
For Use With (Equipment) 8783:Model9044:Spacing6999:English Capacity7679:Speed7681:Controller Type8888:Metric Operating Temperature8887:English Operating Temperature8745:Certifications9955:Electrical Requirements8693:Wattage7680:Voltage8683:Hertz9952:Dimensions (L x W x H)8735:Metric Depth8734:English Depth6967:Metric Length6994:English Length8574:Metric Width8573:English Width8712:Metric Height8711:English Height8855:Metric Shipping Weight8854:English Shipping Weight8689:Metric Mass8882:Humidity Range8741:Quantity6973:For Use With Equipment8540:For Use With Application8560:Format
Quantity 3
Spacing 7 in.
Format 7 in.