Please login to your online account to display your discounted pricing

Wheaton™ Bottom-Drive R2P 2.0 Roller Apparatus


Advanced Production Scale Roller Culture Equipment

Manufacturer: wheaton™  WWRBMR5010A

 View more versions of this product

Catalog No. 02-924-952

  • / Each
 

Description & Specifications

Specifications

Capacity (English) 5 Bottle positions
For Use With (Application) For laboratory, pilot plant and production cell culture installations
Quantity 1
Spacing 7 in.
Format 7 in.