missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ BOD Bottles

Available with either glass robotic or pennyhead stoppers

Manufacturer:  DWK Life Sciences 22749718

 View more versions of this product

Catalog No. 02-926-44


 

Description

Description

Bottles

  • Borosilicate glass, 60 or 300mL, numbered or unnumbered
  • With permanent marking areas; numbered versions are also bar-coded
  • Specially designed shoulder radius sweeps all air from inside the bottle during filling
  • 300mL bottles are recommended for the five-day BOD test referenced in Standard Methods for the Examination of Water and Wastewater, 18th Edition, 5210B, 1992
Stoppers
  • Interchangeable borosilicate glass stoppers have tapered bottoms that also prevent air entrapment
  • Flared mouth of bottle forms a water seal around stoppers that prevents air from being drawn in during incubation
  • Stopper joint is compatible with the probes of the leading BOD and DO meters
  • Glass robotic stoppers are compatible with most robotic systems

Use to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action.

Order Info

Dimensions: 300mL bottles are 2.75 diameter × 6.5 in. high (69 × 165mm); 60mL bottles are 1.75 diameter × 4.5 in. high (43 × 115mm)

Specifications

Specifications

300mL
2.7 in.
6.49 in.
Borosilicate Glass
409 to 432
Robotic
10.14 oz.
6.9cm
16.5cm
Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action
Yes
10.14 oz., 300mL
Documents
Accessories