Please login to your online account to display your discounted pricing

Wheaton™ Black Phenolic Screw Caps with PTFE faced 14B Rubber Liners for E-Z Ex-Traction™ Vials


Designed to ensure the ultimate in product safety

Manufacturer: wheaton™  WW240825

 View more versions of this product

Catalog No. 02-927-105

  • / Case of 500 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/Rubber
Color Black
For Use With 10mL Vial
Neck Finish 22-400
No. per Case 500 Cs.