Please login to your online account to display your discounted pricing

Wheaton™ Black Phenolic Screw Caps with PTFE faced 14B Rubber Liners for E-Z Ex-Traction™ Vials


Designed to ensure the ultimate in product safety

Manufacturer: wheaton™  WW240823

 View more versions of this product

Catalog No. 02-927-103

  • / Case of 1000 Each
 

Description & Specifications

Specifications

For Use With (Equipment) 8mL Vial
Neck Finish 15-425
Color Black
Certifications/Compliance ISO 9001
No. per Case 1000 Cs.
Closure Liner PTFE/Rubber

  • Autoclavable
  • Liner made of styrene-butadiene rubber faced with PTFE
  • Excellent chemical resistance