DWK Life Sciences Wheaton™ Black Phenolic Screw Cap With Rubber Liner
Nontoxic white styrene-butadiene rubber liner
Manufacturer: DWK Life Sciences 240208
Description
- Autoclavable for easy sterilization
Specifications
ISO 9001 | |
Bottle | |
Phenolic | |
13-425 |
14B Rubber | |
Black | |
200 Cs. |