DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with PTFE-Faced Rubber Liner
Features PTFE faced (14B) styrene-butadiene rubber liner
Manufacturer: DWK Life Sciences 240480
Description
- Designed for product safety
- TFE/Styrene-Butadiene rubber liner
- Screw cap
- Pheolic
- Black
- Autoclavable
Specifications
Autoclavable | |
PTFE/14B Rubber | |
Black | |
100 Cs. |
ISO 9001 | |
Bottle | |
Phenolic | |
33-430 |