Please login to your online account to display your discounted pricing

Wheaton™ Black Phenolic Screw Cap with PTFE-Faced Rubber Liner


Features PTFE faced (14B) styrene-butadiene rubber liner

Manufacturer: wheaton™  240414

 View more versions of this product

Catalog No. 02-912-525

  • / Case of 200 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/14B Rubber
Color Black
Autoclavable Yes
For Use With Bottle
Material Phenolic
Neck Finish 18-415
No. per Case 200 Cs.