DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with Metal Foil Liner
Features an aluminum foil faced pulp board liner that can be used with organic solvents
Manufacturer: DWK Life Sciences 240321
Description
- Good resistance to hydrocarbons, oils, ketones and alcohols
- Not good for acids or alkalis
- Closure size: 33mm-400
- Phenolic
- Autoclavable
- Black
Specifications
ISO 9001 | |
Bottle | |
Phenolic | |
33-400 |
Metal Foil | |
Black | |
100 Cs. |