DWK Life Sciences Wheaton™ Black Phenolic Cap with Rubber Liner
14B rubber-lined caps feature a nontoxic, white, styrene-butadiene rubber liner, ideal for cell culture work
Supplier: DWK Life Sciences W239302
Specifications
Rubber | |
Black | |
1000 Cs. | |
ISO 9001 |
Bottle | |
Phenolic Resin | |
33-400 |