Please login to your online account to display your discounted pricing

Wheaton™ Black Phenolic Cap with Rubber Liner


14B rubber-lined caps feature a nontoxic, white, styrene-butadiene rubber liner, ideal for cell culture work

Manufacturer: wheaton™  W239301

 View more versions of this product

Catalog No. 02-912-248

  • / Case of 144 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Bottle
Neck Finish 28-400
Certifications/Compliance ISO 9001
Color Black
No. per Case 1000 Cs.
Closure Liner Rubber