Please login to your online account to display your discounted pricing

Wheaton™ Black Phenolic Cap with Rubber Liner


14B rubber-lined caps feature a nontoxic, white, styrene-butadiene rubber liner, ideal for cell culture work

Manufacturer: wheaton™  W239304

 View more versions of this product

Catalog No. 02-912-251

  • / Case of 72 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Bottle
Neck Finish 43-400
Material Phenolic
Certifications/Compliance ISO 9001
Color Black
Closure Liner Rubber
No. per Case 1000 Cs.