Wheaton™ Biobanking System
Integrated kit offers the Wheaton PluraScan and SingleScan with packaged CryoELITE Vials, KeepIT Boxes and 2D Data Matrix Bar Code Inserts
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
03-410-058 Dwk Life Sciences Inc
W986020
Internal vials, 1 PluraScan, 1 Singlescan, 2 cases 2D data matrix bar code inserts, 2 cases white internal thread CryoELITE™ cryogenic vials, 1 case white KeepIT 100 storage boxes N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-410-059 Dwk Life Sciences Inc
W986030
External vials, 1 PluraScan, 1 Singlescan, 2 cases 2D data matrix bar code inserts, 2 cases white internal thread CryoELITE™ cryogenic vials, 1 case white KeepIT 100 storage boxes N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
The Wheaton 2D Integrated BioBanking System is a comprehensive offering of high-quality products customized to enhance the storage, tracking and management of your vital samples. Whether you require the storage of irreplaceable samples in glass cryogenic ampules or plastic cryogenic vials, bar coding options for sample traceability or color coded storage to track specific projects or samples, Wheaton provides you with the tools that make your job easier and more efficient.
Ampules
  • Cryogenic Wheaton Cryule™ Ampules are ideal for long-term storage integrity due to their tamper evident seal
  • Ampules are not manufactured pre-bar coded, enabling customization of bar codes according to customer specifications
  • For use with scanners, a 2D bar code is applied to the ampule
  • Ceramic bar code is durable when used in extreme temperatures
CryoELITE™ Cryogenic Vials
  • Unrivaled cap seals exceed DOT and IATA regulations
  • Optional 2D Data Matrix Bar Code Inserts provide a unique identifier for traceability
  • Bottom format allows unrestricted view of 2D bar code, convenient for automated scanning
  • Made from low binding, cryogenic grade virgin polypropylene, with lot certified RNase/DNase and Endotoxin-free certification
  • Colored caps allow for color coding projects along with Wheaton color coded KeepIT Boxes
2D Bar Codes
  • When purchasing freestanding Wheaton CryoELITE Vials, you can also get optional 2D Data Matrix Bar Code Inserts that allow for immediate bar coding of samples
  • Insert can also be applied to vial at a future date, which eliminates jeopardizing the integrity of sample by transferring it to another vial
  • 2D Data Matrix Bar Code Insert press fits and locks into place in bottom of vial
  • 2D bar codes are machine readable symbols of rows of encrypted data arranged in a rectangular or square pattern that store large amounts of data
  • Number sequencing on bar codes is guaranteed not to duplicate
KeepIT™ Boxes
  • Provide an ideal method for batching and storing of samples
  • Openings in bottom of box facilitate scanning of either CryoELITE Vials with 2D Data Matrix Bar Code Inserts or ceramic 2D bar code labeled Cryule™ Ampules
SingleScan™ Bar Code Reader
  • Able to read 1D and 2D bar codes
  • Linear bar codes are black and white vertical bars located on side of ampule that contain various amounts of data ranging from simple number chains to large amounts of numeric and alphanumeric data sets
  • 2D bar codes are machine readable symbols of rows of encrypted data arranged in a rectangular or square pattern that store large amounts of data
PluraScan™ Bar Code Reader
  • Able to read 2D bar coded vials or ampules stored in compatible, bottom readable Wheaton KeepIT Boxes
  • Once bar code is attached to ampule or vial, bar codes can be scanned and read using Wheaton SingleScan or PluraScan Bar Code Reader
  • Bar code identification can be simply stored in a Microsoft™ Excel™ file or any other data collection system
Sample Storage, Traceability and Management:
  • Ampules
  • CryoELITE™ Vials
  • 2D Bar Code Inserts
  • KeepIT™ Boxes
  • Scanners
Description & Specifications
Specifications
0.0