Wheaton™ Amber Wide-Mouth Packers with Poly-Seal™-lined Closures
Amber glass for storing light-sensitive powders
$60.06 - $89.43
Specifications
Closure Liner Polyseal
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Amber
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity Closure Size Diameter (English) Height (English) Price Quantity  
02-912-120 Wheaton™
W216952
2 oz., 60mL 33-400 1.73 in. 3.03 in. Case of 24 Each for $64.83
02-912-148 Wheaton™
W216951
1 oz., 30mL 28-400 1.45 in. 2.63 in. Case of 24 Each for $60.06
02-912-149 Wheaton™
W216953
4 oz., 125mL 38-400 2.12 in. 3.81 in. Case of 24 Each for $89.43
Specifications
Description
  • USP Type III soda-lime glass
  • Allows for easy removal of contents
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Polyseal
Screw
Amber
Phenolic
Black
Soda Lime Glass