Please login to your online account to display your discounted pricing

Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper


Ideal for shelf storage

Manufacturer: wheaton™  W215255

 View more versions of this product

Catalog No. 02-911-306

  • / Case of 6 Each
 

Description & Specifications

Specifications

Closure Color Amber
Height (Metric) 115mm
Diameter (Metric) 53mm
Mouth Narrow
Capacity (English) 3.38 oz.
Material Glass
Diameter (English) 2.08 in.
Color Amber
Capacity (Metric) 100mL
Capacity 3.38 oz., 100mL
Closure Material Borosilicate Glass
Height (English) 4.52 in.