Please login to your online account to display your discounted pricing

Wheaton™ Amber Glass Reagent Bottle with Ground Glass Stopper


Ideal for shelf storage

Manufacturer: wheaton™  215259

 View more versions of this product

Catalog No. 02-918B

  • / Case of 10 Each
 

Description & Specifications

Specifications

Capacity 16.9 oz., 500mL
Capacity (Metric) 500mL
Capacity (English) 16.9 oz.
Closure Material Borosilicate Glass
Color Amber
Diameter (English) 3.46 in.
Height (English) 6.88 in.
Closure Color Amber
Diameter (Metric) 88 mm
Height (Metric) 175mm
Mouth Narrow